(3S)-2-Carbobenzoxy-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: Z-Tic-OH, (S)-N-Benzyloxycarbonyl-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, (3S)-2-Cbz-1,2,3,4-tetrahydroisoquinoline-3-carboxylic Acid, Z-L-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, (3S)-2-[(Benzyloxy)carbonyl]-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid

A275497-100G A275497-10G A275497-1G A275497-25G
77289-530EA 176.13 USD
77289-530 77289-532 77289-534 77289-536
(3S)-2-Carbobenzoxy-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid 97%
(3S)-2-Carbobenzoxy-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₈H₁₇NO₄
MW: 311.3 g/mol
MDL Number: MFCD00672545
CAS Number: 79261-58-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR