4-Isopropylcyclohexanone 98%

Supplier: Ambeed
Warning

A275427-100MG A275427-1G A275427-250MG A275427-25G A275427-5G
77289-520EA 6 USD
77289-520 77289-522 77289-524 77289-526 77289-528
4-Isopropylcyclohexanone 98%
4-Isopropylcyclohexanone

Precaución: for Research Use Only

Formula: C₉H₁₆O
MDL Number: MFCD00043479
CAS Number: 5432-85-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR