3-Nitroquinolin-2-ol 95%

Supplier: Ambeed
Warning

A275293-100MG A275293-250MG
77238-806EA 27 USD
77238-806 77238-808
3-Nitroquinolin-2-ol 95%
3-Nitroquinolin-2-ol

Precaución: for Research Use Only

Formula: C₉H₆N₂O₃
MDL Number: MFCD09754108
CAS Number: 103029-75-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR