Piperidin-3-amine dihydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: piperidin-3-amine dihydrochloride, 3-Aminopiperidine dihydrochloride, 3-Amino-Piperidine Di-Hydrochloride

A275279-100G A275279-1G A275279-25G A275279-5G
77289-512EA 239 USD
77289-512 77289-514 77289-516 77289-518
Piperidin-3-amine dihydrochloride 95%
Piperidin-3-amine dihydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₄Cl₂N₂
MW: 173.081 g/mol
MDL Number: MFCD00012773
CAS Number: 138060-07-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR