4-Isopropyl-m-cresol 98%

Supplier: Ambeed
Warning

A275269-10G A275269-25G A275269-500G A275269-5G
77289-506EA 8 USD
77289-506 77289-508 77289-510 77571-696
4-Isopropyl-m-cresol 98%
4-Isopropyl-m-cresol

Precaución: for Research Use Only

Formula: C₁₀H₁₄O
MDL Number: MFCD00010704
CAS Number: 3228-02-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR