2,4,5-Trimethylaniline 98%

Supplier: Ambeed
Danger

A275235-100MG A275235-10G A275235-1G A275235-250MG A275235-5G
77250-404EA 25.75 USD
77250-404 77250-406 77250-408 77250-410 77250-412
2,4,5-Trimethylaniline 98%
2,4,5-Trimethylaniline

Precaución: for Research Use Only

Formula: C₉H₁₃N
MW: 135.21 g/mol
MDL Number: MFCD00025201
CAS Number: 137-17-7
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR