2-Bromo-4-fluorobenzyl alcohol 98%

Supplier: Ambeed
Warning

Synonyms: 2-Bromo-4-fluorobenzyl Alcohol, (2-Bromo-4-fluorophenyl)methanol

A275193-100G A275193-25G A275193-5G
77262-544EA 112.27 USD
77262-544 77262-546 77262-548
2-Bromo-4-fluorobenzyl alcohol 98%
2-Bromo-4-fluorobenzyl alcohol

Precaución: for Research Use Only

Formula: C₇H₆BrFO
MW: 205.03 g/mol
MDL Number: MFCD00672925
CAS Number: 229027-89-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR