1-Azabicyclo[2.2.2]octane-4-carbonitrile 97%

Supplier: Ambeed
Danger

Synonyms: 4-Cyanonquinuclidine, Quinuclidine-4-carbonitrile, 4-Cyano-1-azabicyclo[2.2.2]octane

A274566-250MG A274566-10G A274566-5G
77289-492EA 41.2 USD
77289-492 77616-152 77748-526
1-Azabicyclo[2.2.2]octane-4-carbonitrile 97%
1-Azabicyclo[2.2.2]octane-4-carbonitrile

Precaución: for Research Use Only

Formula: C₈H₁₂N₂
MW: 136.194 g/mol
MDL Number: MFCD00276610
CAS Number: 26458-78-6
UN: 3439
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR