2-Chloro-3-iodo-5-(trifluoromethyl)pyridine 95%

Supplier: Ambeed
Danger

A274551-1G A274551-250MG
77262-540EA 182.31 USD
77262-540 77262-542
2-Chloro-3-iodo-5-(trifluoromethyl)pyridine 95%
2-Chloro-3-iodo-5-(trifluoromethyl)pyridine

Precaución: for Research Use Only

Formula: C₆H₂ClF₃IN
MDL Number: MFCD08741352
CAS Number: 505084-56-0
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR