4-Bromo-N-phenylaniline 98%

Supplier: Ambeed
Danger

A274297-25G A274297-5G A274297-1G
77289-488EA 21 USD
77289-488 77289-490 77748-525
4-Bromo-N-phenylaniline 98%
4-Bromo-N-phenylaniline

Precaución: for Research Use Only

Formula: C₁₂H₁₀BrN
MDL Number: MFCD07779504
CAS Number: 54446-36-5
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR