(+)-Di-p-toluoyl-D-tartaric acid 98%

Supplier: Ambeed
Warning

Synonyms: Di-p-toluoyl-D-tartaric acid, (2S,3S)-(+)-Di-O-4-Toluoyl-D-Tartaric Acid, Di-p-toluoyl-D-tartaric Acid

A274172-1000G A274172-100G A274172-25G A274172-500G A274172-5G
77289-478EA 63 USD
77289-478 77289-480 77289-482 77289-484 77289-486
(+)-Di-p-toluoyl-D-tartaric acid 98%
(+)-Di-p-toluoyl-D-tartaric acid

Precaución: for Research Use Only

Formula: C₂₀H₁₈O₈
MDL Number: MFCD00008552
CAS Number: 32634-68-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR