2-Bromo-5-chloro-4-fluoroaniline 97%

Supplier: Ambeed
Warning

A273919-100G A273919-5G
77262-536EA 562.99 USD
77262-536 77262-538
2-Bromo-5-chloro-4-fluoroaniline 97%
2-Bromo-5-chloro-4-fluoroaniline

Precaución: for Research Use Only

Formula: C₆H₄BrClFN
MDL Number: MFCD10698796
CAS Number: 85462-59-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR