3,4-Dimethoxyphenylacetone 97%

Supplier: Ambeed
Warning

Synonyms: 1-(3,4-Dimethoxyphenyl)propan-2-one

A273831-1000G A273831-100G A273831-10G A273831-25G A273831-500G A273831-5G
77276-826EA 247.2 USD
77276-826 77276-828 77276-830 77276-832 77276-834 77276-836
3,4-Dimethoxyphenylacetone 97%
3,4-Dimethoxyphenylacetone

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₃
MW: 194.23 g/mol
MDL Number: MFCD00008772
CAS Number: 776-99-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR