1-Methyl-1H-1,3-benzimidazole-5-carboxylic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-Methyl-1H-benzo[d]imidazole-5-carboxylic acid
A273651-100MG
A273651-1G
A273651-250MG
A273651-25G
A273651-5G
77238-786EA
18.54
USD
77238-786
77238-788
77238-790
77238-792
77238-794
1-Methyl-1H-1,3-benzimidazole-5-carboxylic acid 98%
1-Methyl-1H-1,3-benzimidazole-5-carboxylic acid
Precaución: for Research Use Only
Formula:
C₉H₈N₂O₂ MW: 176.18 g/mol |
MDL Number:
MFCD06809665 CAS Number: 53484-17-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...