Sodium 4,4'-oxybis(butane-1-sulfonate) 95%

Supplier: Ambeed
Warning

Synonyms: Bis(4-sulphobutyl)ether Disodium Salt

A273611-1G A273611-250MG A273611-5G A273611-100MG
77238-784EA 297.67 USD
77238-784 77601-198 77601-200 77746-484
Sodium 4,4'-oxybis(butane-1-sulfonate) 95%
Sodium 4,4'-oxybis(butane-1-sulfonate)

Precaución: for Research Use Only

Formula: C₈H₁₆Na₂O₇S₂
MW: 334.32 g/mol
MDL Number: MFCD28138233
CAS Number: 183278-30-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR