Bay-K-8644 (S)-(-)- 98%

Supplier: Ambeed
Warning

Synonyms: Methyl (S)-2,6-dimethyl-5-nitro-4-(2-(trifluoromethyl)phenyl)-1,4-dihydropyridine-3-carboxylate, (S)-(-)-Bay-K-8644

A273444-10MG A273444-25MG
77250-390EA 64.89 USD
77250-390 77250-392
Bay-K-8644 (S)-(-)- 98%
Bay-K-8644 (S)-(-)-

Precaución: for Research Use Only

Formula: C₁₆H₁₅F₃N₂O₄
MW: 356.30 g/mol
MDL Number: MFCD00153769
CAS Number: 98625-26-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR