1-Boc-indole-5-boronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole-1-carboxylate

A273223-1G A273223-5G
77289-468EA 36 USD
77289-468 77289-470
1-Boc-indole-5-boronic acid pinacol ester 98%
1-Boc-indole-5-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₉H₂₆BNO₄
MW: 343.2251 g/mol
MDL Number: MFCD05663895
CAS Number: 777061-36-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR