(S)-2-Amino-2-(3-ethoxy-4-methoxyphenyl)acetic acid 97%

Supplier: Ambeed
Warning

A273125-1G A273125-250MG
77262-518EA 41.2 USD
77262-518 77262-520
(S)-2-Amino-2-(3-ethoxy-4-methoxyphenyl)acetic acid 97%
(S)-2-Amino-2-(3-ethoxy-4-methoxyphenyl)acetic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₅NO₄
MW: 225.24 g/mol
MDL Number: MFCD14572374
CAS Number: 1212977-59-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR