3,3'-Diamino diphenyl sulfone 97%

Supplier: Ambeed
Warning

Synonyms: 3,3'-Sulphonyldianiline, 3,3'-sulphonyldianiline, 3-Aminophenyl sulphone, DADPS

A273084-1000G A273084-100G A273084-10G A273084-25G A273084-500G
77276-816EA 63.86 USD
77276-816 77276-818 77276-820 77276-822 77276-824
3,3'-Diamino diphenyl sulfone 97%
3,3'-Diamino diphenyl sulphone

Precaución: for Research Use Only

Formula: C₁₂H₁₂N₂O₂S
MW: 248.31 g/mol
MDL Number: MFCD00007792
CAS Number: 599-61-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR