2-Imidazol-1-ylethylamine 95%

Supplier: Ambeed
Danger

Synonyms: 2-(1H-Imidazol-1-yl)ethanamine

A273074-250MG A273074-5G A273074-1G
77262-514EA 50.47 USD
77262-514 77262-516 77550-694
2-Imidazol-1-ylethylamine 95%
2-Imidazol-1-ylethylamine

Precaución: for Research Use Only

Formula: C₅H₉N₃
MW: 111.15 g/mol
MDL Number: MFCD03093042
CAS Number: 5739-10-6
UN: 2735
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR