2-Biphenylyl isocyanate 98%

Supplier: Ambeed
Danger

A273013-1G A273013-25G A273013-5G
77262-508EA 21 USD
77262-508 77262-510 77262-512
2-Biphenylyl isocyanate 98%
2-Biphenylyl isocyanate

Precaución: for Research Use Only

Formula: C₁₃H₉NO
MDL Number: MFCD00037087
CAS Number: 17337-13-2
UN: 2810
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR