2-(2-(2-(2-((Tetrahydro-2H-pyran-2-yl)oxy)ethoxy)ethoxy)ethoxy)ethyl 4-methylbenzenesulfonate 97%

Supplier: Ambeed
Warning

Synonyms: Tos-PEG4-THP

A272990-1G A272990-250MG
77262-504EA 76.22 USD
77262-504 77262-506
2-(2-(2-(2-((Tetrahydro-2H-pyran-2-yl)oxy)ethoxy)ethoxy)ethoxy)ethyl 4-methylbenzenesulfonate 97%
2-(2-(2-(2-((Tetrahydro-2H-pyran-2-yl)oxy)ethoxy)ethoxy)ethoxy)ethyl 4-methylbenzenesulfonate

Precaución: for Research Use Only

Formula: C₂₀H₃₂O₈S
MW: 432.52 g/mol
MDL Number: MFCD28976701
CAS Number: 86259-89-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR