2-Amino-2-[4-(trifluoromethyl)phenyl]acetic acid 98%

Supplier: Ambeed
Warning

Synonyms: H-DL-Phg(4-CF₃)-OH, 4-(Trifluoromethyl)-DL-phenylglycine, 2-Amino-2-(4-(trifluoromethyl)phenyl)acetic acid

A272851-100MG A272851-1G A272851-250MG
77262-498EA 28.84 USD
77262-498 77262-500 77262-502
2-Amino-2-[4-(trifluoromethyl)phenyl]acetic acid 98%
2-Amino-2-[4-(trifluoromethyl)phenyl]acetic acid

Precaución: for Research Use Only

Formula: C₉H₈F₃NO₂
MW: 219.16 g/mol
MDL Number: MFCD00665333
CAS Number: 142012-65-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR