2-Chloro-4-nitrobenzotrifluoride 98%

Supplier: Ambeed
Warning

A272529-100G A272529-10G A272529-1G A272529-25G A272529-5G
77262-486EA 258 USD
77262-486 77262-488 77262-490 77262-492 77262-494
2-Chloro-4-nitrobenzotrifluoride 98%
2-Chloro-4-nitrobenzotrifluoride

Precaución: for Research Use Only

Formula: C₇H₃ClF₃NO₂
MDL Number: MFCD00171532
CAS Number: 151504-80-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR