N-(Chloroacetyl)morpholine 97%

Supplier: Ambeed
Danger

Synonyms: 4-(Chloroacetyl)morpholine, 4-(2-Chloroacetyl)morpholine

A272158-25G A272158-5G A272158-10G
77289-464EA 41 USD
77289-464 77289-466 77593-678
N-(Chloroacetyl)morpholine 97%
N-(Chloroacetyl)morpholine

Precaución: for Research Use Only

Formula: C₆H₁₀ClNO₂
MDL Number: MFCD00721939
CAS Number: 1440-61-5
UN: 3265
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR