N,N-Dimethyl-1H-benzo[d]imidazol-2-amine 97%

Supplier: Ambeed
Warning

A272098-1G A272098-250MG A272098-5G
77238-770EA 216.3 USD
77238-770 77238-772 77238-774
N,N-Dimethyl-1H-benzo[d]imidazol-2-amine 97%
N,N-Dimethyl-1H-benzo[d]imidazol-2-amine

Precaución: for Research Use Only

Formula: C₉H₁₁N₃
MW: 161.20 g/mol
MDL Number: MFCD01231011
CAS Number: 2851-13-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR