trans,trans-4'-Pentyl-4-(3,4,5-trifluorophenyl)bicyclohexyl 97%

Supplier: Ambeed
Warning

Synonyms: (trans,trans)-4-Pentyl-4'-(3,4,5-trifluorophenyl)-1,1'-bi(cyclohexane), trans,trans-4'-Amyl-4-(3,4,5-trifluorophenyl)bicyclohexyl

A272056-1G A272056-5G
77250-380EA 7.21 USD
77250-380 77250-382
trans,trans-4'-Pentyl-4-(3,4,5-trifluorophenyl)bicyclohexyl 97%
trans,trans-4'-Pentyl-4-(3,4,5-trifluorophenyl)bicyclohexyl

Precaución: for Research Use Only

Formula: C₂₃H₃₃F₃
MW: 366.51 g/mol
MDL Number: MFCD17019208
CAS Number: 137644-54-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR