3,3'-Diaminobenzophenone 95%

Supplier: Ambeed
Warning

A271991-1G A271991-25G A271991-5G
77276-788EA 36 USD
77276-788 77276-790 77748-521
3,3'-Diaminobenzophenone 95%
3,3'-Diaminobenzophenone

Precaución: for Research Use Only

Formula: C₁₃H₁₂N₂O
MDL Number: MFCD00014774
CAS Number: 611-79-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR