4-(1-Piperidinylmethyl)-phenol 97%

Supplier: Ambeed
Warning

Synonyms: 4-(Piperidin-1-ylmethyl)phenol

A271926-100MG A271926-1G A271926-250MG
77238-764EA 44 USD
77238-764 77238-766 77238-768
4-(1-Piperidinylmethyl)-phenol 97%
4-(1-Piperidinylmethyl)-phenol

Precaución: for Research Use Only

Formula: C₁₂H₁₇NO
MDL Number: MFCD18453499
CAS Number: 73152-41-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR