BRD7116 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
N,N'-(sulphonylbis(4,1-phenylene))bis(2,2,3,3-tetramethylcyclopropane-1-carboxamide), N,N'-(sulphonyldi-4,1-phenylene)bis[2,2,3,3-tetramethylcyclopropanecarboxamide]
A271536-10MG
A271536-1MG
A271536-25MG
A271536-50MG
A271536-5MG
77238-740EA
14.42
USD
77238-740
77238-742
77238-744
77238-746
77238-748
BRD7116 98%
BRD7116
Precaución: for Research Use Only
Formula:
C₂₈H₃₆N₂O₄S MW: 496.66 g/mol |
MDL Number:
MFCD02168947 CAS Number: 329059-55-4 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...