(2,6-Difluorophenyl)acetic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2,6-Difluorophenylacetic acid

A271428-100G A271428-10G A271428-1G A271428-25G A271428-5G
77238-732EA 231.75 USD
77238-732 77238-734 77238-736 77238-738 77616-148
(2,6-Difluorophenyl)acetic acid 97%
(2,6-Difluorophenyl)acetic acid

Precaución: for Research Use Only

Formula: (C₆H₃F₂)CH₂COOH
MW: 172.13 g/mol
MDL Number: MFCD00010001
CAS Number: 85068-28-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR