9,9-Di-n-hexylfluorene-2,7-diboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: (9,9-Dihexyl-9H-fluorene-2,7-diyl)diboronic acid

A271347-1G A271347-250MG A271347-5G
77250-360EA 30 USD
77250-360 77250-362 77250-364
9,9-Di-n-hexylfluorene-2,7-diboronic acid 97%
9,9-Di-n-hexylfluorene-2,7-diboronic acid

Precaución: for Research Use Only

Formula: C₂₅H₃₆B₂O₄
MDL Number: MFCD03427279
CAS Number: 203927-98-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR