trans-4'-(p-Tolyl)-[1,1'-bi(cyclohexan)]-4-one 97%

Supplier: Ambeed
Warning

Synonyms: trans-4'-(4-Methylphenyl)[1,1'-bi(cyclohexyl)]-4-one, (1'r,4'r)-4'-(p-Tolyl)-[1,1'-bi(cyclohexan)]-4-one, trans-4'-(p-tolyl)-[1,1'-Bi(cyclohexan)]-4-one

A271292-100MG A271292-1G
77238-728EA 5 USD
77238-728 77238-730
trans-4'-(p-Tolyl)-[1,1'-bi(cyclohexan)]-4-one 97%
trans-4'-(p-Tolyl)-[1,1'-bi(cyclohexan)]-4-one

Precaución: for Research Use Only

Formula: C₁₉H₂₆O
MW: 270.42 g/mol
MDL Number: MFCD13194928
CAS Number: 125962-80-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR