2-Methylbiphenyl 98%

Supplier: Ambeed
Warning

Synonyms: 2-Phenyltoluene

A270970-1G A270970-5G
77276-770EA 16.48 USD
77276-770 77276-772
2-Methylbiphenyl 98%
2-Methylbiphenyl

Precaución: for Research Use Only

Formula: C₁₃H₁₂
MW: 168.24 g/mol
MDL Number: MFCD00008517
CAS Number: 643-58-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR