6-Amino-9-((2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-7H-purine-8(9H)-thione 97%

Supplier: Ambeed
Warning

Synonyms: 8-Mercaptoadenosine

A270946-100MG A270946-1G A270946-250MG
77250-338EA 12.36 USD
77250-338 77250-340 77250-342
6-Amino-9-((2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-7H-purine-8(9H)-thione 97%
6-Amino-9-((2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-7H-purine-8(9H)-thione

Precaución: for Research Use Only

Formula: C₁₀H₁₃N₅O₄S
MW: 299.30 g/mol
CAS Number: 3001-45-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR