N'-(4-Amino-2-cyanophenyl)-N,N-dimethylformimidamide 97%

Supplier: Ambeed
Warning

A270800-100MG A270800-1G A270800-250MG
77262-452EA 5 USD
77262-452 77262-454 77262-456
N'-(4-Amino-2-cyanophenyl)-N,N-dimethylformimidamide 97%
N'-(4-Amino-2-cyanophenyl)-N,N-dimethylformimidamide

Precaución: for Research Use Only

Formula: C₁₀H₁₂N₄
MDL Number: MFCD09998252
CAS Number: 194423-17-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR