2-Methoxybiphenyl 98%

Supplier: Ambeed
Warning

Synonyms: Biphenyl-2-yl methyl ether, 2-Phenylanisole

A270433-100G A270433-25G A270433-5G
77262-440EA 189.52 USD
77262-440 77262-442 77262-444
2-Methoxybiphenyl 98%
2-Methoxybiphenyl

Precaución: for Research Use Only

Formula: C₁₃H₁₂O
MW: 184.24 g/mol
MDL Number: MFCD00008367
CAS Number: 86-26-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR