1H-[1,2,3]Triazolo[4,5-c]pyridine 98%

Supplier: Ambeed
Warning

Synonyms: 3H-[1,2,3]Triazolo[4,5-c]pyridine

A270421-100MG A270421-10G A270421-1G A270421-250MG A270421-25G A270421-5G
77238-712EA 5 USD
77238-712 77238-714 77238-716 77238-718 77238-720 77238-722
1H-[1,2,3]Triazolo[4,5-c]pyridine 98%
1H-[1,2,3]Triazolo[4,5-c]pyridine

Precaución: for Research Use Only

Formula: C₅H₄N₄
MW: 120.112 g/mol
MDL Number: MFCD02670924
CAS Number: 273-05-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR