4,6-Dimethylnicotinonitrile 97%

Supplier: Ambeed
Warning

A270228-1G A270228-250MG A270228-25G A270228-5G
77289-426EA 25 USD
77289-426 77289-428 77289-430 77289-432
4,6-Dimethylnicotinonitrile 97%
4,6-Dimethylnicotinonitrile

Precaución: for Research Use Only

Formula: C₈H₈N₂
MDL Number: MFCD00234419
CAS Number: 6623-21-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR