3-Cyclohexene-1-methanol 98%

Supplier: Ambeed
Warning

Synonyms: 1,2,3,6-Tetrahydrobenzyl Alcohol, Cyclohex-3-en-1-ylmethanol

A270197-1000G A270197-100G A270197-25G A270197-500G A270197-5G
77276-754EA 200.85 USD
77276-754 77276-756 77276-758 77276-760 77276-762
3-Cyclohexene-1-methanol 98%
3-Cyclohexene-1-methanol

Precaución: for Research Use Only

Formula: C₇H₁₂O
MW: 112.17 g/mol
MDL Number: MFCD00001574
CAS Number: 1679-51-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR