5-Methylthiophene-2-carboxaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: 5-Methylthiophene-2-Carbaldehyde, 5-Methyl-2-thiophenecarboxaldehyde, 5-Methylthiophene-2-carbaldehyde

A270093-10G A270093-25G A270093-5G A270093-100G
77289-412EA 9 USD
77289-412 77289-414 77289-416 77616-140
5-Methylthiophene-2-carboxaldehyde 98%
5-Methylthiophene-2-carboxaldehyde

Precaución: for Research Use Only

Formula: C₆H₆OS
MDL Number: MFCD00005434
CAS Number: 13679-70-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR