Ethyl 2-((2-ethoxyphenyl)amino)-2-oxoacetate 97%

Supplier: Ambeed
Danger

A269969-1G A269969-250MG A269969-5G
77238-704EA 47.38 USD
77238-704 77238-706 77238-708
Ethyl 2-((2-ethoxyphenyl)amino)-2-oxoacetate 97%
Ethyl 2-((2-ethoxyphenyl)amino)-2-oxoacetate

Precaución: for Research Use Only

Formula: C₁₂H₁₅NO₄
MW: 237.25 g/mol
MDL Number: MFCD01839248
CAS Number: 24439-50-7
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR