1,4-Bis(4-nitrophenyl)butane-1,4-dione 97%

Supplier: Ambeed
Warning

A269843-1G A269843-250MG A269843-5G
77262-424EA 67 USD
77262-424 77262-426 77262-428
1,4-Bis(4-nitrophenyl)butane-1,4-dione 97%
1,4-Bis(4-nitrophenyl)butane-1,4-dione

Precaución: for Research Use Only

Formula: C₁₆H₁₂N₂O₆
MW: 328.27 g/mol
MDL Number: MFCD20482624
CAS Number: 108791-66-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR