6-Bromo-7-fluoroquinazolin-4-ol 98%

Supplier: Ambeed
Warning

A269639-100MG A269639-1G A269639-250MG
77238-698EA 29.87 USD
77238-698 77238-700 77238-702
6-Bromo-7-fluoroquinazolin-4-ol 98%
6-Bromo-7-fluoroquinazolin-4-ol

Precaución: for Research Use Only

Formula: C₈H₄BrFN₂O
MW: 243.03 g/mol
MDL Number: MFCD09907649
CAS Number: 950577-02-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR