1,1-Bis[(2S,5S)-2,5-diethylphospholano]ferrocene ≥98%, ee 99%

Supplier: Ambeed
Warning

A269623-100MG A269623-250MG A269623-5G A269623-10G A269623-1G
77250-320EA 37 USD
77250-320 77250-322 77250-324 77746-468 77746-469
1,1-Bis[(2S,5S)-2,5-diethylphospholano]ferrocene ≥98%, ee 99%
1,1-Bis[(2S,5S)-2,5-diethylphospholano]ferrocene

Precaución: for Research Use Only

Formula: C₂₆H₄₀FeP₂
MW: 470.388 g/mol
MDL Number: MFCD09265150
CAS Number: 436863-50-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR