2,6-Dichloroquinoline 97%

Supplier: Ambeed
Warning

A269610-100G A269610-10G A269610-1G A269610-250MG A269610-25G A269610-5G
77250-308EA 457.01 USD
77250-308 77250-310 77250-312 77250-314 77250-316 77250-318
2,6-Dichloroquinoline 97%
2,6-Dichloroquinoline

Precaución: for Research Use Only

Formula: C₉H₅Cl₂N
MDL Number: MFCD03427194
CAS Number: 1810-72-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR