1-Bromo-2-ethoxy-4-nitrobenzene 97%

Supplier: Ambeed
Warning

Synonyms: 2-Bromo-5-nitrophenetole

A269526-10G A269526-1G A269526-250MG
77262-418EA 47.38 USD
77262-418 77262-420 77262-422
1-Bromo-2-ethoxy-4-nitrobenzene 97%
1-Bromo-2-ethoxy-4-nitrobenzene

Precaución: for Research Use Only

Formula: C₈H₈BrNO₃
MW: 246.06 g/mol
MDL Number: MFCD03790890
CAS Number: 423165-33-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR