CZC-25146 98%

Supplier: Ambeed
Warning

Synonyms: N-(2-((5-Fluoro-2-((2-methoxy-4-morpholinophenyl)amino)pyrimidin-4-yl)amino)phenyl)methanesulphonamide

A269266-100MG A269266-10MG A269266-1G A269266-1MG A269266-5MG
77238-688EA 164.8 USD
77238-688 77238-690 77238-692 77238-694 77238-696
CZC-25146 98%
CZC-25146

Precaución: for Research Use Only

Formula: C₂₂H₂₅FN₆O₄S
MW: 488.54 g/mol
MDL Number: MFCD28160475
CAS Number: 1191911-26-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR