2,5-Dichloro-4-nitroaniline 98%
Supplier: Ambeed
|
Warning
|
A269232-10G
A269232-1G
A269232-25G
A269232-5G
77289-398EA
51.5
USD
77289-398
77289-400
77289-402
77289-404
2,5-Dichloro-4-nitroaniline 98%
2,5-Dichloro-4-nitroaniline
Precaución: for Research Use Only
Formula:
C₆H₄Cl₂N₂O₂ MW: 207.02 g/mol |
MDL Number:
MFCD00025157 CAS Number: 6627-34-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...