1'-Benzyl-6-chloro-3H-spiro[furo[3,4-c]pyridine-1,4'-piperidin]-3-one 97%

Supplier: Ambeed
Warning

A269210-1G A269210-250MG
77262-414EA 119.48 USD
77262-414 77262-416
1'-Benzyl-6-chloro-3H-spiro[furo[3,4-c]pyridine-1,4'-piperidin]-3-one 97%
1'-Benzyl-6-chloro-3H-spiro[furo[3,4-c]pyridine-1,4'-piperidin]-3-one

Precaución: for Research Use Only

Formula: C₁₈H₁₇ClN₂O₂
MW: 328.79 g/mol
MDL Number: MFCD30730081
CAS Number: 1017598-71-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR